This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01319239_m1
See other MTRF1 GE Assays ›
Gene Symbol:
Mtrf1
Gene Name:
mitochondrial translational release factor 1
Gene Aliases:
A830062K05Rik, MtRF-1
Chromosome Location:
Chr.14: 79397761 - 79423650 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01319239_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mtrf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145960.4 NP_666072.3 9 - 10 1509 57
GenBank mRNA AK043981.1 - 9 - 10 1315 57
AK051834.1 - 9 - 10 1295 57
AK077749.1 - 9 - 10 1510 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001102.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mtrf1

Panther Classification:

Molecular Function -

translation release factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational termination
molecular_function
translation release factor activity
translation release factor activity, codon specific

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400008808.mm.1 NM_145960.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400008808.mm.1 NM_145960.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400008808.mm.1 TC1400001102.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0