This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01319272_m1
See other NEK2 GE Assays ›
Gene Symbol:
Nek2
Gene Name:
NIMA (never in mitosis gene a)-related expressed kinase 2
Gene Aliases:
AA617254, C77054
Chromosome Location:
Chr.1: 191820875 - 191833049 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01319272_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nek2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010892.3 NP_035022.2 3 - 4 723 73
GenBank mRNA AF007247.1 - 3 - 4 637 73
AF013166.1 - 3 - 4 607 73
AK077627.1 - 3 - 4 687 73
AK147072.1 - 3 - 4 699 73
AK164467.1 - 3 - 4 666 73
AK169330.1 - 4 - 5 614 73
AK170179.1 - 3 - 4 643 73
AK171277.1 - 3 - 4 643 73
AK171387.1 - 3 - 4 643 73
BC010302.1 - 3 - 4 645 3597227 73
BC057576.1 - 3 - 4 663 6810914 73
U95610.1 - 3 - 4 634 73

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nek2

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

mitotic sister chromatid segregation
blastocyst development
protein phosphorylation
cell cycle
chromosome segregation
mitotic nuclear division
phosphorylation
negative regulation of DNA binding
protein autophosphorylation
centrosome separation
cell division
meiotic cell cycle
regulation of attachment of spindle microtubules to kinetochore
mitotic spindle assembly
negative regulation of centriole-centriole cohesion
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein phosphatase binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0