This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01319675_mH
See other RARB GE Assays ›
Gene Symbol:
Rarb
Gene Name:
retinoic acid receptor, beta
Gene Aliases:
A830025K23, Hap, Nr1b2
Chromosome Location:
Chr.14: 16430839 - 17082331 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01319675_mH
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rarb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289760.1 NP_001276689.1 4 - 5 883 88
NM_001289761.1 NP_001276690.1 3 - 4 802 88
NM_001289762.1 NP_001276691.1 2 - 3 412 88
NM_011243.2 NP_035373.1 2 - 3 766 88
GenBank mRNA AK043726.1 - 2 - 3 89
AK078471.1 - 2 - 3 479 88
BC076597.1 - 2 - 3 762 30608242 88
S56660.1 - 2 - 3 662 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001487.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Nuclear Receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rarb

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
ureteric bud development
glandular epithelial cell development
outflow tract septum morphogenesis
retinal pigment epithelium development
growth plate cartilage development
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of cell proliferation
negative regulation of cell proliferation
striatum development
neurogenesis
regulation of myelination
negative regulation of chondrocyte differentiation
embryonic hindlimb morphogenesis
multicellular organism growth
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of programmed cell death
steroid hormone mediated signaling pathway
positive regulation of neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
embryonic eye morphogenesis
retinoic acid receptor signaling pathway
embryonic digestive tract development
ventricular cardiac muscle cell differentiation
retina development in camera-type eye
bone development
negative regulation of cartilage development
RNA polymerase II regulatory region sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
retinoic acid receptor activity
drug binding
zinc ion binding
protein complex binding
sequence-specific DNA binding
metal ion binding
retinoid X receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400011137.mm.1 NM_001289760.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400011137.mm.1 NM_001289761.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400011137.mm.1 NM_001289762.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400011137.mm.1 NM_011243.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400011137.mm.1 TC1400001487.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400011142.mm.1 NM_001289762.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400011142.mm.1 NM_011243.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400011142.mm.1 TC1400001487.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400011143.mm.1 NM_001289762.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400011143.mm.1 NM_011243.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400011143.mm.1 TC1400001487.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0