Assay Details
| Gene Symbol: Mef2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291191.1 | NP_001278120.1 | 5 - 6 | 749 | 146 | |
| NM_001291192.1 | NP_001278121.1 | 5 - 6 | 749 | 146 | ||
| GenBank mRNA | AK040227.1 | - | 1 - 2 | 645 | 145 | |
| AK153751.1 | - | 5 - 6 | 750 | 146 | ||
| BC096598.1 | - | 5 - 6 | 691 | 4979487 | 146 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mef2a
Genomic Map