This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01320034_g1
See other SLC7A7 GE Assays ›
Gene Symbol:
Slc7a7
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 7
Gene Aliases:
AI790233, my+lat1
Chromosome Location:
Chr.14: 54369442 - 54417702 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01320034_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253680.1 NP_001240609.1 1 - 2 223 78
GenBank mRNA AJ012754.1 - 1 - 2 213 78
AK152637.1 - 1 - 2 128 78
AK170851.1 - 1 - 2 134 78
BC014709.1 - 1 - 2 211 4482816 78
CB953352.1 - 1 - 2 226 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AJ012754.1, AK152637.1, AK170851.1, BC014709.1, NM_001253680.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

regulation of arginine metabolic process
amino acid transmembrane transport
transport
amino acid transport
basic amino acid transport
L-alpha-amino acid transmembrane transport
amino acid transmembrane transporter activity
basic amino acid transmembrane transporter activity
L-amino acid transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400015426.mm.1 NM_001253679.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015426.mm.1 NM_001253679.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015426.mm.1 NM_001253680.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015426.mm.1 NM_001253680.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015426.mm.1 NM_011405.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015426.mm.1 NM_011405.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400015426.mm.1 TC1400002054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400015427.mm.1 NM_001253680.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015427.mm.1 NM_001253680.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015427.mm.1 TC1400002054.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 728261 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0