This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01320035_m1
See other SLC7A7 GE Assays ›
Gene Symbol:
Slc7a7
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 7
Gene Aliases:
AI790233, my+lat1
Chromosome Location:
Chr.14: 54369442 - 54417702 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01320035_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253679.1 NP_001240608.1 3 - 4 748 87
NM_001253680.1 NP_001240609.1 2 - 3 766 87
NM_011405.4 NP_035535.2 4 - 5 905 87
GenBank mRNA AJ012754.1 - 2 - 3 756 87
AJ130943.1 - 3 - 4 748 87
AK150950.1 - 4 - 5 905 87
AK151718.1 - 4 - 5 905 87
AK152637.1 - 2 - 3 671 87
AK170851.1 - 2 - 3 677 87
BC014709.1 - 2 - 3 754 4482816 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002054.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a7

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

regulation of arginine metabolic process
amino acid transmembrane transport
transport
amino acid transport
basic amino acid transport
L-alpha-amino acid transmembrane transport
amino acid transmembrane transporter activity
basic amino acid transmembrane transporter activity
L-amino acid transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400015430.mm.1 NM_001253679.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015430.mm.1 NM_001253679.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015430.mm.1 NM_011405.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015430.mm.1 NM_011405.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400015430.mm.1 TC1400002054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400015425.mm.1 NM_001253679.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015425.mm.1 NM_001253679.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015425.mm.1 NM_001253680.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015425.mm.1 NM_001253680.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015425.mm.1 NM_011405.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015425.mm.1 NM_011405.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400015425.mm.1 TC1400002054.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400015429.mm.1 NM_011405.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015429.mm.1 NM_011405.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400015429.mm.1 TC1400002054.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0