This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01320315_m1
See other PPA2 GE Assays ›
Gene Symbol:
Ppa2
Gene Name:
pyrophosphatase (inorganic) 2
Gene Aliases:
1110013G13Rik, Sid6306
Chromosome Location:
Chr.3: 133309159 - 133378235 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01320315_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppa2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001293641.1 NP_001280570.1 3 - 4 276 94
NM_146141.2 NP_666253.1 3 - 4 276 94
GenBank mRNA AA444705.1 - 3 - 4 275 94
AK090384.1 - 3 - 4 265 94
AK143475.1 - 3 - 4 247 94
BC011417.1 - 3 - 4 270 2654305 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001354.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppa2

Panther Classification:

Molecular Function -

pyrophosphatase

Gene Ontology Categories:

Function(s) Process(es)

phosphate-containing compound metabolic process
dephosphorylation
magnesium ion binding
inorganic diphosphatase activity
protein serine/threonine phosphatase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300010876.mm.1 NM_146141.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300010876.mm.1 TC0300001354.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0