This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01320489_g1
See other FURIN GE Assays ›
Gene Symbol:
Furin
Gene Name:
furin (paired basic amino acid cleaving enzyme)
Gene Aliases:
9130404I01Rik, Fur, PACE, Pcsk3, SPC1
Chromosome Location:
Chr.7: 80389194 - 80405445 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01320489_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Furin
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081454.1 NP_001074923.1 7 - 8 936 66
NM_011046.2 NP_035176.1 7 - 8 1056 66
GenBank mRNA BC048234.1 - 6 - 7 763 6492009 66
L26489.1 - 7 - 8 1056 66
X54056.1 - 6 - 7 759 66

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Furin

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

signal peptide processing
proteolysis
cell proliferation
regulation of signal transduction
negative regulation of endopeptidase activity
protein processing
peptide hormone processing
positive regulation of cell migration
positive regulation of transforming growth factor beta receptor signaling pathway
negative regulation of low-density lipoprotein particle receptor catabolic process
nerve growth factor production
negative regulation of transforming growth factor beta1 production
secretion by cell
regulation of protein catabolic process
peptide biosynthetic process
regulation of low-density lipoprotein particle receptor biosynthetic process
regulation of endopeptidase activity
protease binding
endopeptidase activity
serine-type endopeptidase activity
serine-type endopeptidase inhibitor activity
protein binding
peptidase activity
serine-type peptidase activity
hydrolase activity
peptide binding
metal ion binding
nerve growth factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0