This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01320613_m1
See other PACRG GE Assays ›
Gene Symbol:
Pacrg
Gene Name:
PARK2 co-regulated
Gene Aliases:
1700008H23Rik
Chromosome Location:
Chr.17: 10403012 - 10840191 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01320613_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pacrg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027032.2 NP_081308.1 3 - 4 733 83
GenBank mRNA AK005771.1 - 3 - 4 729 83
AK135510.1 - 3 - 4 735 83
BC120740.1 - 3 - 4 711 40129663 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001439.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pacrg

Gene Ontology Categories:

Function(s) Process(es)

spermatid development
cellular response to unfolded protein
negative regulation of cell death
G-protein coupled receptor binding
actin binding
Hsp70 protein binding
heat shock protein binding
ubiquitin protein ligase binding
alpha-tubulin binding
beta-tubulin binding
chaperone binding
Hsp90 protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700012816.mm.1 NM_027032.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700012816.mm.1 NM_027032.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700012816.mm.1 TC1700001439.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0