Assay Details
| Gene Symbol: Ndst4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022565.2 | NP_072087.2 | 7 - 8 | 2373 | 87 | |
| GenBank mRNA | AB036838.1 | - | 6 - 7 | 1823 | 87 | |
| AK132938.1 | - | 2 - 3 | 528 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndst4
Genomic Map