This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01320969_g1
See other VCAM1 GE Assays ›
Gene Symbol:
Vcam1
Gene Name:
vascular cell adhesion molecule 1
Gene Aliases:
CD106, Vcam-1
Chromosome Location:
Chr.3: 116110020 - 116129688 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01320969_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vcam1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011693.3 NP_035823.3 1 - 2 384 97
GenBank mRNA AK016465.1 - 1 - 2 154 97
AK030195.1 - 1 - 2 154 97
AK143401.1 - 1 - 2 154 97
AK162954.1 - 1 - 2 154 97

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Vcam1

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
leukocyte cell-cell adhesion
cell-matrix adhesion
heart development
amine metabolic process
viral process
single organismal cell-cell adhesion
positive regulation of T cell proliferation
leukocyte tethering or rolling
oxidation-reduction process
cell chemotaxis
embryonic placenta morphogenesis
chorio-allantoic fusion
visceral serous pericardium development
cellular response to glucose stimulus
integrin binding
primary amine oxidase activity
cell adhesion molecule binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0