This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01321140_m1
See other LATS2 GE Assays ›
Gene Symbol:
Lats2
Gene Name:
large tumor suppressor 2
Gene Aliases:
4932411G09Rik, AV277261, AW228608
Chromosome Location:
Chr.14: 57689662 - 57746123 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01321140_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lats2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015771.2 NP_056586.2 5 - 6 2797 70
GenBank mRNA AB023958.1 - 4 - 5 2494 70
AK029242.1 - 2778 70
AK029870.1 - 5 - 6 2778 70
AK029966.1 - 5 - 6 2757 70
AK081190.2 - 6 - 7 3049 70
AK161243.1 - 4 - 5 2219 70
AK163607.1 - 5 - 6 2780 70
AK164789.1 - 5 - 6 2750 70
BC053028.1 - 5 - 6 2751 6409832 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002868.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lats2

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
protein phosphorylation
cell cycle
mitotic nuclear division
hormone-mediated signaling pathway
phosphorylation
keratinocyte differentiation
cellular protein localization
hippo signaling
intracellular signal transduction
negative regulation of cyclin-dependent protein serine/threonine kinase activity
regulation of organ growth
cell division
negative regulation of canonical Wnt signaling pathway
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400016556.mm.1 NM_015771.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016556.mm.1 NM_015771.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400016556.mm.1 TC1400002868.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline