This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01321211_m1
See other GMPR2 GE Assays ›
Gene Symbol:
Gmpr2
Gene Name:
guanosine monophosphate reductase 2
Gene Aliases:
1810008P16Rik, 5730544D12Rik, AA959850
Chromosome Location:
Chr.14: 55672235 - 55687634 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01321211_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gmpr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177992.2 NP_818773.1 8 - 9 856 97
GenBank mRNA AK031004.1 - 8 - 9 858 97
AK075772.1 - 8 - 9 814 97
BC003886.1 - 8 - 9 815 3590411 97
BC024109.1 - 8 - 9 812 5149353 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400000790.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gmpr2

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

purine nucleobase metabolic process
nucleotide metabolic process
monocyte differentiation
GMP catabolic process
oxidation-reduction process
catalytic activity
GMP reductase activity
oxidoreductase activity
metal ion binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430327 BC003886.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430327 BC024109.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430327 AK031004.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430327 AK075772.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430327 NM_177992.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430327 BC003886.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430327 BC024109.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430327 AK031004.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430327 AK075772.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P430327 NM_177992.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621153 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 680520 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300590 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111790 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 140987 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430327 BC003886.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430327 BC024109.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430327 AK031004.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430327 AK075772.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430327 NM_177992.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430327 BC003886.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430327 BC024109.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430327 AK031004.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430327 AK075772.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P430327 NM_177992.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430327 BC003886.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430327 BC024109.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430327 AK031004.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430327 AK075772.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430327 NM_177992.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430327 BC003886.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430327 BC024109.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430327 AK031004.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430327 AK075772.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430327 NM_177992.2 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08119220 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166467 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08119212 - -
Whole Mouse Genome G4122A A_51_P430327 BC003886.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P430327 BC024109.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P430327 AK031004.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P430327 AK075772.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P430327 NM_177992.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P430327 BC003886.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P430327 BC024109.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P430327 AK031004.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P430327 AK075772.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P430327 NM_177992.2 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166467 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11784143 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11784149 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11784145 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11195324 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619279 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619221 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619294 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619249 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619260 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619272 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619241 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619239 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619304 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619215 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619238 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619295 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619291 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619237 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619210 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619269 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619283 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619227 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619255 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619302 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619286 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619252 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619300 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619288 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619280 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619290 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619265 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619232 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619236 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619257 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619247 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619226 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619234 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619208 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619218 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619213 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619277 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619244 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619224 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619250 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619219 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619262 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619285 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619251 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29619245 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619222 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11129949 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619253 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619207 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619213 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619236 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619260 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11784145 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619238 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619259 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619219 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619241 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619229 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619215 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619239 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619218 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619257 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619249 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619245 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619226 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619232 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619224 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619237 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619265 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619244 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619250 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11768146 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619252 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619255 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11195340 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619234 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619227 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619264 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11129948 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619210 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11129951 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29619208 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10415371 - -
Affymetrix Mouse Gene 1.0 ST Array 10415367 - -
Affymetrix Mouse Gene 1.0 ST Array 10415365 - -
Affymetrix Mouse Gene 1.0 ST Array 10415370 - -
Affymetrix Mouse Gene 1.0 ST Array 10415373 - -
Affymetrix Mouse Gene 1.0 ST Array 10415368 - -
Affymetrix Mouse Gene 1.0 ST Array 10415369 - -
Affymetrix Mouse Gene 1.0 ST Array 10415364 - -
Affymetrix Mouse Gene 1.0 ST Array 10415366 - -
Affymetrix Mouse Gene 1.0 ST Array 10415372 - -
Affymetrix Mouse Gene 1.0 ST Array 10415375 - -
Affymetrix HT MG-430 PM Array Plate 1416356_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1429180_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10415363 - -
Mouse Genome 430 2.0 Array 1429180_at - -
Mouse Genome 430 2.0 Array 1416356_PM_at - -
Mouse Genome 430 2.0 Array 1429180_PM_at - -
Mouse Genome 430 2.0 Array 1416356_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 105446_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 105446_at - -
Affymetrix Mouse Clariom S Array PSR1400006422.mm.1 NM_177992.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400006422.mm.1 NM_177992.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400006422.mm.1 TC1400000790.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400006421.mm.1 NM_177992.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400006421.mm.1 NM_177992.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400006421.mm.1 TC1400000790.mm.1 MTA TC
Mouse Expression Set 430 1416356_PM_at - -
Mouse Expression Set 430 1416356_at - -
Mouse Expression Set 430 1429180_at - -
Mouse Expression Set 430 1429180_PM_at - -
Murine Genome U74v2 Set 139141_s_at - -
Murine Genome U74v2 Set 139142_r_at - -
Murine Genome U74v2 Set 139141_PM_s_at - -
Murine Genome U74v2 Set 139142_PM_r_at - -
Mouse Genome 430A 2.0 Array 1416356_PM_at - -
Mouse Genome 430A 2.0 Array 1416356_at - -
Affymetrix HT Mouse Genome 430A Array 1416356_at - -
Affymetrix HT Mouse Genome 430A Array 1416356_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1429180_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1429180_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 4920204 - -
Illumina mouse-6 v1.0 expression beadchip 4920204 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004920204 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2617691 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2617691 - -
mouse-6 v1.1 (Illumina) scl000307.1_98-S - -
Illumina Sentrix mouse-6.v1 bead array 4920204 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl000307.1_98-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4920204 - -
MouseRef-8 v2 Expression BeadChip ILMN_2846317 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2617691 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2846317 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline