This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01322606_m1
See other SOCS4 GE Assays ›
Gene Symbol:
Socs4
Gene Name:
suppressor of cytokine signaling 4
Gene Aliases:
3110032M18Rik, A730004F22Rik, AI427843, AU015727, AU015859, Socs7
Chromosome Location:
Chr.14: 47277143 - 47291591 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01322606_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Socs4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080843.2 NP_543119.2 1 - 2 81 96
GenBank mRNA AK014115.1 - 1 - 2 112 96
BC057159.1 - 1 - 2 212 96

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK014115.1, BC057159.1, NM_080843.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Socs4

Panther Classification:

Molecular Function -

kinase modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein kinase activity
negative regulation of epidermal growth factor-activated receptor activity
negative regulation of signal transduction
cytokine-mediated signaling pathway
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
intracellular signal transduction
regulation of growth
negative regulation of epidermal growth factor receptor signaling pathway
negative regulation of JAK-STAT cascade
molecular_function
protein kinase inhibitor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400004136.mm.1 NM_080843.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400004136.mm.1 NM_080843.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400004136.mm.1 TC1400000510.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400004137.mm.1 NM_080843.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400004137.mm.1 NM_080843.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400004137.mm.1 TC1400000510.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline