This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01323309_m1
See other PLD5 GE Assays ›
Gene Symbol:
Pld5
Gene Name:
phospholipase D family, member 5
Gene Aliases:
B230365F16Rik
Chromosome Location:
Chr.1: 175962301 - 176280017 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01323309_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pld5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195816.1 NP_001182745.1 1 - 2 329 60
GenBank mRNA AK082725.1 - 1 - 2 229 60
BC100428.1 - 1 - 2 72 30247182 60
BU609501.1 - 1 - 2 298 60
CK626039.1 - 1 - 2 329 60

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC100428.1, NM_001195816.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
catalytic activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100029712.mm.1 NM_001195816.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029712.mm.1 NM_001195816.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029712.mm.1 TC0100003614.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100029710.mm.1 NM_001195816.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029710.mm.1 NM_001195816.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029710.mm.1 TC0100003614.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100029711.mm.1 NM_001195816.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029711.mm.1 NM_001195816.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029711.mm.1 TC0100003614.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100029709.mm.1 NM_001195816.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029709.mm.1 NM_001195816.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029709.mm.1 NM_176916.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029709.mm.1 NM_176916.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100029709.mm.1 TC0100003614.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 330705 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0