This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01323313_m1
See other PLD5 GE Assays ›
Gene Symbol:
Pld5
Gene Name:
phospholipase D family, member 5
Gene Aliases:
B230365F16Rik
Chromosome Location:
Chr.1: 175962301 - 176280017 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01323313_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pld5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195816.1 NP_001182745.1 6 - 7 1065 87
NM_176916.4 NP_795890.2 6 - 7 1369 87
GenBank mRNA AK136851.1 - 5 - 6 1183 87
AK136888.1 - 5 - 6 1183 87
AK144116.1 - 6 - 7 87
BC100428.1 - 6 - 7 30247182 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003614.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pld5

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
catalytic activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100029700.mm.1 NM_001195816.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029700.mm.1 NM_001195816.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029700.mm.1 NM_176916.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029700.mm.1 NM_176916.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100029700.mm.1 TC0100003614.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0