This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01323361_mH
See other ACADM GE Assays ›
Gene Symbol:
Acadm
Gene Name:
acyl-Coenzyme A dehydrogenase, medium chain
Gene Aliases:
AU018656, MCAD
Chromosome Location:
Chr.3: 153922353 - 153944643 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01323361_mH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acadm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007382.5 NP_031408.1 6 - 7 687 101
GenBank mRNA AK131793.1 - 6 - 7 489 101
AK146444.1 - 6 - 7 492 101
BC013498.1 - 6 - 7 472 4222450 101
U07159.1 - 6 - 7 476 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300003173.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acadm

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

liver development
glycogen biosynthetic process
organic acid metabolic process
regulation of gluconeogenesis
lipid metabolic process
fatty acid metabolic process
fatty acid beta-oxidation
heart development
metabolic process
response to cold
carnitine metabolic process
post-embryonic development
carnitine metabolic process, CoA-linked
fatty acid beta-oxidation using acyl-CoA dehydrogenase
response to starvation
carnitine biosynthetic process
protein homotetramerization
medium-chain fatty acid metabolic process
medium-chain fatty acid catabolic process
cardiac muscle cell differentiation
oxidation-reduction process
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
isomerase activity
identical protein binding
flavin adenine dinucleotide binding
medium-chain-acyl-CoA dehydrogenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0