This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01323635_m1
See other FIRRE GE Assays ›
Gene Symbol:
Firre
Gene Name:
functional intergenic repeating RNA element
Gene Aliases:
5830467J12Rik, 6720401G13Rik, AW048145
Chromosome Location:
Chr.X: 50555744 - 50635258 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01323635_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Firre
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK081581.1 - 4 - 5 1510 108
KJ638686.1 - 10 - 11 2400 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P223508 CO044012.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P223508 EL608124.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P223508 NR_026976.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P223508 AK030946.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P223508 AK081581.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P223508 CO044012.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P223508 EL608124.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P223508 NR_026976.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P223508 AK030946.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P223508 AK081581.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181934 BC055934.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181934 BC066100.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181934 NR_015505.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072193 BC066100.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072193 NR_026976.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072193 AK081581.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072193 NR_015505.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 379258 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 116438 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537215 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499294 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802070 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537574 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952853 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 106034 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 118412 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 747042 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839640 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785798 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348418 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 64245 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 641538 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 64439 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 1887 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490926 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 671426 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 374943 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 383796 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 672894 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 322391 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 895231 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242024 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828235 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67542 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864906 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198908 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651439 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793873 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 480602 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614388 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191850 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226379 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198420 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69603 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628607 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94198 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495997 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 148010 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110355 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24858 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 90469 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34861 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16271 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 10442 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 10660 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181934 BC055934.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181934 BC066100.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181934 NR_015505.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072193 BC066100.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072193 NR_026976.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072193 AK081581.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072193 NR_015505.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P223508 CO044012.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P223508 EL608124.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P223508 NR_026976.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P223508 AK030946.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P223508 AK081581.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P223508 CO044012.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P223508 EL608124.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P223508 NR_026976.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P223508 AK030946.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P223508 AK081581.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P290152 BP762728.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P290152 BC055934.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P290152 BC066100.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P290152 AK032694.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P290152 NR_015505.1 mouse_transcriptome
Agilent Six_ChIP_03_of_03_v2 A_68_P32372305 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14832108 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14880319 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14972881 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372332 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372319 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372262 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372126 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372301 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372299 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372551 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372590 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372081 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372296 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372298 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14832109 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372256 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372107 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15166648 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372128 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372263 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372587 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372584 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372557 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15008090 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372267 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14862703 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372286 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372105 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14972890 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372261 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372258 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372259 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14796903 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372336 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372120 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14664656 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14862697 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372084 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372124 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372565 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372579 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372552 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372102 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372137 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372308 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372586 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372593 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372293 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372564 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15189455 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372558 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372311 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372583 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372257 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372585 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15076287 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372303 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372333 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14988656 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15074403 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372556 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372268 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372283 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372580 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372567 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372096 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372251 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372302 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372078 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372561 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372569 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15166637 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15074404 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15166643 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15023259 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372122 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372307 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372592 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372085 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14988652 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372264 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372341 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372345 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372589 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372130 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15166635 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15189454 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372594 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372576 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372273 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372288 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14796907 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372294 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372274 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372560 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372570 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372553 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372276 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P15008104 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14796896 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372588 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32372309 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10604507 - -
Affymetrix Mouse Gene 1.0 ST Array 10604506 - -
Affymetrix HT MG-430 PM Array Plate 1438916_PM_x_at - -
Affymetrix HT MG-430 PM Array Plate 1435744_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1436638_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1438915_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10604505 - -
Mouse Genome 430 2.0 Array 1436638_at - -
Mouse Genome 430 2.0 Array 1438915_at - -
Mouse Genome 430 2.0 Array 1438915_PM_at - -
Mouse Genome 430 2.0 Array 1436638_PM_at - -
Mouse Genome 430 2.0 Array 1435744_PM_at - -
Mouse Genome 430 2.0 Array 1438916_x_at - -
Mouse Genome 430 2.0 Array 1438916_PM_x_at - -
Mouse Genome 430 2.0 Array 1435744_at - -
Mouse Expression Set 430 1438916_PM_x_at - -
Mouse Expression Set 430 1436638_at - -
Mouse Expression Set 430 1438915_at - -
Mouse Expression Set 430 1435744_at - -
Mouse Expression Set 430 1436638_PM_at - -
Mouse Expression Set 430 1438916_x_at - -
Mouse Expression Set 430 1438915_PM_at - -
Mouse Expression Set 430 1435744_PM_at - -
Murine Genome U74v2 Set 166734_PM_at - -
Murine Genome U74v2 Set 166734_at - -
Affymetrix HT Mouse Genome 430B Array 1436638_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1438915_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1436638_at - -
Affymetrix HT Mouse Genome 430B Array 1435744_at - -
Affymetrix HT Mouse Genome 430B Array 1438916_PM_x_at - -
Affymetrix HT Mouse Genome 430B Array 1438915_at - -
Affymetrix HT Mouse Genome 430B Array 1438916_x_at - -
Affymetrix HT Mouse Genome 430B Array 1435744_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 714990 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline