This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01323797_m1
See other ACIN1 GE Assays ›
Gene Symbol:
Acin1
Gene Name:
apoptotic chromatin condensation inducer 1
Gene Aliases:
2610036I19Rik, 2610510L13Rik, Acinus, Acn, C79325, acinusL, acinusS, mKIAA0670
Chromosome Location:
Chr.14: 54642161 - 54686931 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01323797_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acin1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001085473.2 NP_001078942.2 7 - 8 2332 62
NM_023190.3 NP_075679.2 7 - 8 2332 62
GenBank mRNA AK030449.1 - 7 - 8 2025 62
AK050467.1 - 6 - 7 1895 62
AK122342.1 - 7 - 8 2003 62
BC075691.1 - 7 - 8 2017 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002063.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acin1

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
mRNA processing
apoptotic process
RNA splicing
regulation of autophagy
apoptotic chromosome condensation
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of mRNA splicing, via spliceosome
mitophagy in response to mitochondrial depolarization
nucleotide binding
nucleic acid binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400015578.mm.1 NM_001085473.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015578.mm.1 NM_001085473.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400015578.mm.1 NM_023190.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015578.mm.1 NM_023190.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400015578.mm.1 TC1400002063.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline