This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01323961_m1
See other IL16 GE Assays ›
Gene Symbol:
Il16
Gene Name:
interleukin 16
Gene Aliases:
mKIAA4048
Chromosome Location:
Chr.7: 83643063 - 83746271 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01323961_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010551.3 NP_034681.2 2 - 3 508 89
GenBank mRNA AF175292.1 - 2 - 3 508 89
AK137237.1 - 2 - 3 467 89
AK220455.1 - 2 - 3 518 89
BC058709.1 - 2 - 3 541 6841305 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003641.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il16

Panther Classification:

Molecular Function -

interleukin superfamily

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
chemotaxis
leukocyte chemotaxis
induction of positive chemotaxis
cytokine activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700030951.mm.1 NM_010551.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700030951.mm.1 NM_010551.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700030951.mm.1 TC0700003641.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline