This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01324068_g1
See other IGSF1 GE Assays ›
Gene Symbol:
Igsf1
Gene Name:
immunoglobulin superfamily, member 1
Gene Aliases:
5330413N23, 5530402E03, AI747649, InhBP/p120, mKIAA0364
Chromosome Location:
Chr.X: 49782536 - 49797906 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01324068_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igsf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177591.4 NP_808259.2 17 - 18 3598 68
NM_183336.2 NP_899179.1 8 - 9 1852 68
GenBank mRNA AK030701.1 - 8 - 9 1854 68
AK076427.1 - 8 - 9 1845 68
AY227771.1 - 17 - 18 3592 68
AY227774.1 - 8 - 9 1847 68
BC119264.1 - 8 - 9 1815 8734013 68
BC119266.1 - 8 - 9 1815 8734015 68
BC144762.1 - 8 - 9 1815 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002177.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
signal transduction
receptor activity
coreceptor activity
inhibin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00013828.mm.1 NM_177591.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00013828.mm.1 NM_177591.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00013828.mm.1 NM_183336.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00013828.mm.1 NM_183336.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00013828.mm.1 TC0X00002177.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6s775:80/100.66.79.102:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0