This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01324249_m1
See other NAA16 GE Assays ›
Gene Symbol:
Naa16
Gene Name:
N(alpha)-acetyltransferase 16, NatA auxiliary subunit
Gene Aliases:
1300019C06Rik, Narg1l
Chromosome Location:
Chr.14: 79334507 - 79390777 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01324249_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Naa16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025832.2 NP_080108.1 14 - 15 1804 98
GenBank mRNA AK005056.1 - 14 - 15 1814 98
AK007755.1 - 14 - 15 1840 98
AK028432.1 - 14 - 15 1915 98
AK152498.1 - 14 - 15 1814 98
BC048727.1 - 2 - 3 345 98
BC052445.1 - 14 - 15 1804 6412745 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002490.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Naa16

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

N-terminal protein amino acid acetylation
negative regulation of apoptotic process
protein stabilization
peptide alpha-N-acetyltransferase activity
ribosome binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400019126.mm.1 NM_025832.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019126.mm.1 NM_025832.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400019126.mm.1 TC1400002490.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline