This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01324595_g1
See other NDUFAF1 GE Assays ›
Gene Symbol:
Ndufaf1
Gene Name:
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 1
Gene Aliases:
2410001M24Rik, CGI-65, CIA30
Chromosome Location:
Chr.2: 119655446 - 119662885 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01324595_g1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufaf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027175.3 NP_081451.3 4 - 5 1104 80
GenBank mRNA AK010328.1 - 3 - 4 934 80
AK050343.1 - 4 - 5 1105 80
BC018422.1 - 4 - 5 934 3593650 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004443.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufaf1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
NADH dehydrogenase (ubiquinone) activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21570807 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21570806 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157922 NM_027175.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157922 AK010328.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157922 AK050343.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2157922 BC018422.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194267 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 675986 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 745310 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513393 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77345 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177369 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157922 NM_027175.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157922 AK010328.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157922 AK050343.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2157922 BC018422.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176854 NM_027175.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176854 AK010328.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176854 AK050343.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P176854 BC018422.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570804 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570813 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570810 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570800 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570795 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570831 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570796 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570793 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570821 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570820 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570818 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570834 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570809 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570814 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570797 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570807 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570806 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570811 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570822 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21570802 - -
Whole Mouse Genome G4122A A_51_P176854 NM_027175.3 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P176854 AK010328.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P176854 AK050343.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P176854 BC018422.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109028 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01180848 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01197425 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01273788 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01273789 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21570807 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21570806 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570791 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570806 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570757 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570847 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570820 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570795 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570780 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570760 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570846 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570851 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570800 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570768 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570818 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570789 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570776 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570772 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570821 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570814 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570853 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570807 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570815 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570803 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570762 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570850 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570767 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570857 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570811 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570783 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570831 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570813 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570770 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570777 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570809 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21570810 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570823 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570846 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570837 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01148976 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01386658 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570834 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01460461 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01855989 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570792 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01622261 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02093453 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570783 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570850 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570772 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01197425 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01460464 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01603312 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570763 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570795 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01603314 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570797 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570855 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570786 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01622262 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01561579 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570767 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570853 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570775 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570813 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02251468 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01483077 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570821 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570765 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570800 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01339513 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01273787 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01209843 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01561576 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01197424 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570809 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570803 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570796 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570791 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570770 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570768 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02103913 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570822 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01209852 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570845 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01969567 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01603316 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570787 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21570814 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10486287 - -
Affymetrix Mouse Gene 1.0 ST Array 10486288 - -
Affymetrix Mouse Gene 1.0 ST Array 10486289 - -
Affymetrix Mouse Gene 1.0 ST Array 10486285 - -
Affymetrix Mouse Gene 1.0 ST Array 10486286 - -
Affymetrix HT MG-430 PM Array Plate 1423711_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10486284 - -
Mouse Genome 430 2.0 Array 1423711_PM_at - -
Mouse Genome 430 2.0 Array 1423711_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 69702_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 69702_PM_at - -
Affymetrix Mouse Clariom S Array PSR0200037099.mm.1 NM_027175.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037099.mm.1 NM_027175.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200037099.mm.1 TC0200004443.mm.1 MTA TC
Mouse Expression Set 430 1423711_at - -
Mouse Expression Set 430 1423711_PM_at - -
Murine Genome U74v2 Set 161507_f_at - -
Murine Genome U74v2 Set 161507_PM_f_at - -
Murine Genome U74v2 Set 100892_at - -
Murine Genome U74v2 Set 100892_PM_at - -
Mouse Genome 430A 2.0 Array 1423711_PM_at - -
Mouse Genome 430A 2.0 Array 1423711_at - -
Affymetrix HT Mouse Genome 430A Array 1423711_at - -
Affymetrix HT Mouse Genome 430A Array 1423711_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 919327 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 130113 - -
Illumina mouse-6 v1.0 expression beadchip 130113 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000130113 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248304 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1248304 - -
mouse-6 v1.1 (Illumina) scl18802.5.1_208-S - -
Illumina Sentrix mouse-6.v1 bead array 130113 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl18802.5.1_208-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 130113 - -
MouseRef-8 v2 Expression BeadChip ILMN_1248304 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248304 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl18802.5.1_208-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline