This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01325774_g1
See other PPP2R5C GE Assays ›
Gene Symbol:
Ppp2r5c
Gene Name:
protein phosphatase 2, regulatory subunit B', gamma
Gene Aliases:
2610043M05Rik, 2700063L20Rik, AI060890, AW545884, C85228, D12Bwg0916e, mKIAA0044
Chromosome Location:
Chr.12: 110447179 - 110583075 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01325774_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r5c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081457.2 NP_001074926.1 4 - 5 615 94
NM_001081458.2 NP_001074927.1 4 - 5 615 94
NM_001135001.1 NP_001128473.1 6 - 7 846 94
NM_012023.3 NP_036153.2 4 - 5 615 94
GenBank mRNA AB055635.1 - 4 - 5 465 94
AB055636.1 - 4 - 5 465 94
AK031584.1 - 4 - 5 618 94
AK129042.1 - 6 - 7 718 94
BC003979.1 - 4 - 5 575 3489999 94
CV558022.1 - 4 - 5 615 94
U37353.1 - 4 - 5 444 94
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK012612.1 - Chr. - - [-] on Build 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001207.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp2r5c

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
proteasome-mediated ubiquitin-dependent protein catabolic process
protein binding
protein phosphatase type 2A regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200008441.mm.1 NM_001081457.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008441.mm.1 NM_001081458.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008441.mm.1 NM_001135001.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200008441.mm.1 NM_001135001.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200008441.mm.1 NM_012023.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008441.mm.1 TC1200001207.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline