This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01326512_g1
See other PLEKHA8 GE Assays ›
Gene Symbol:
Plekha8
Gene Name:
pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 8
Gene Aliases:
6330400G01, AA517676, BC052360, Fapp-2, Fapp2
Chromosome Location:
Chr.6: 54595111 - 54645823 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01326512_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plekha8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001001335.2 NP_001001335.1 12 - 13 1527 112
NM_001164361.1 NP_001157833.1 12 - 13 1622 112
GenBank mRNA AK078040.1 - 12 - 13 1623 112
AK170592.1 - 12 - 13 1387 112
BC052360.1 - 12 - 13 1527 6704789 112

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Plekha8

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

transport
lipid transport
ER to Golgi ceramide transport
glycolipid transport
lipid binding
glycolipid transporter activity
glycolipid binding
phosphatidylinositol-4-phosphate binding
ceramide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600005026.mm.1 NM_001001335.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005026.mm.1 NM_001001335.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600005026.mm.1 NM_001164361.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005026.mm.1 NM_001164361.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005026.mm.1 TC0600000655.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline