| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600005022.mm.1 |
NM_001001335.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005022.mm.1 |
NM_001001335.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005022.mm.1 |
NM_001164361.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005022.mm.1 |
NM_001164361.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005022.mm.1 |
TC0600000655.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005023.mm.1 |
NM_001001335.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005023.mm.1 |
NM_001001335.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005023.mm.1 |
NM_001164361.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005023.mm.1 |
NM_001164361.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005023.mm.1 |
TC0600000655.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600005021.mm.1 |
NM_001001335.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005021.mm.1 |
NM_001001335.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005021.mm.1 |
NM_001164361.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005021.mm.1 |
NM_001164361.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600005021.mm.1 |
TC0600000655.mm.1 |
MTA TC |