This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01326651_g1
See other ZC3H14 GE Assays ›
Gene Symbol:
Zc3h14
Gene Name:
zinc finger CCCH type containing 14
Gene Aliases:
1010001P15Rik, 1700016A15Rik, 2700069A02Rik, AU014748
Chromosome Location:
Chr.12: 98746968 - 98787774 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01326651_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zc3h14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008506.2 NP_001008506.2 6 - 7 1022 84
NM_001160107.1 NP_001153579.1 6 - 7 1022 84
NM_029334.2 NP_083610.2 6 - 7 1022 84
NR_027648.1 5 - 6 907 84
GenBank mRNA AK048046.1 - 6 - 7 988 84
AK050226.1 - 6 - 7 1003 84
AK076066.1 - 5 - 6 903 84
AK160937.1 - 6 - 7 1008 84
AK193300.1 - 1 - 2 161 84
AK196806.1 - 1 - 2 161 84
AK197684.1 - 1 - 2 161 84
AK202592.1 - 1 - 2 161 84
AK202852.1 - 1 - 2 162 84
AK207425.1 - 1 - 2 161 84
AK210490.1 - 1 - 2 161 84
BC021797.1 - 2 - 3 454 84
BC024824.1 - 6 - 7 992 5355748 84
BC026610.1 - 2 - 3 487 84
BC034760.1 - 6 - 7 955 84

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zc3h14

Gene Ontology Categories:

Function(s) Process(es)

biological_process
regulation of mRNA stability
negative regulation of mRNA polyadenylation
RNA binding
poly(A) binding
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200006786.mm.1 NM_001008506.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006786.mm.1 NM_001008506.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006786.mm.1 NM_001160107.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006786.mm.1 NM_001160107.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006786.mm.1 NM_029334.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006786.mm.1 NM_029334.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006786.mm.1 NR_027648.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006786.mm.1 NR_027648.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006786.mm.1 TC1200000970.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200006788.mm.1 NM_001008506.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006788.mm.1 NM_001008506.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006788.mm.1 NM_001160107.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006788.mm.1 NM_001160107.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006788.mm.1 NM_029334.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006788.mm.1 NM_029334.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006788.mm.1 TC1200000970.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200006790.mm.1 NM_001008506.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006790.mm.1 NM_001008506.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006790.mm.1 NM_001160107.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006790.mm.1 NM_001160107.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006790.mm.1 NM_029334.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006790.mm.1 NM_029334.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006790.mm.1 NR_027648.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006790.mm.1 NR_027648.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006790.mm.1 TC1200000970.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200006787.mm.1 NM_001008506.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006787.mm.1 NM_001008506.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006787.mm.1 NM_001160107.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006787.mm.1 NM_001160107.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006787.mm.1 NM_029334.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006787.mm.1 NM_029334.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006787.mm.1 NR_027648.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006787.mm.1 NR_027648.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006787.mm.1 TC1200000970.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200006791.mm.1 NM_001008506.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006791.mm.1 NM_001008506.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006791.mm.1 NM_001160107.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006791.mm.1 NM_001160107.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006791.mm.1 NM_029334.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006791.mm.1 NM_029334.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006791.mm.1 NR_027648.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006791.mm.1 NR_027648.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006791.mm.1 TC1200000970.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200006785.mm.1 NM_001008506.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006785.mm.1 NM_001008506.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006785.mm.1 NM_001160107.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006785.mm.1 NM_001160107.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006785.mm.1 NM_029334.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006785.mm.1 NM_029334.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006785.mm.1 NR_027648.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006785.mm.1 NR_027648.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006785.mm.1 TC1200000970.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200006789.mm.1 NM_001008506.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006789.mm.1 NM_001008506.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006789.mm.1 NM_001160107.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006789.mm.1 NM_001160107.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006789.mm.1 NM_029334.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006789.mm.1 NM_029334.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006789.mm.1 NR_027648.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006789.mm.1 NR_027648.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006789.mm.1 TC1200000970.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200006792.mm.1 NM_001008506.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006792.mm.1 NM_001008506.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006792.mm.1 NM_001160107.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006792.mm.1 NM_001160107.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006792.mm.1 NM_029334.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006792.mm.1 NM_029334.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200006792.mm.1 NR_027648.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006792.mm.1 NR_027648.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200006792.mm.1 TC1200000970.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 660582 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0