This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01326759_m1
See other AGPAT2 GE Assays ›
Gene Symbol:
Agpat2
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 2 (lysophosphatidic acid acyltransferase, beta)
Gene Aliases:
2510002J07Rik, AV000834, BSCL, BSCL1, LPAAB, LPAAT-beta
Chromosome Location:
Chr.2: 26593057 - 26604417 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01326759_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Agpat2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026212.2 NP_080488.1 1 - 2 392 57
GenBank mRNA AK010891.1 - 1 - 2 201 57
AY072769.1 - 1 - 2 185 57
BC125530.1 - 1 - 2 266 40130045 57
BC137840.1 - 1 - 2 266 8860859 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003195.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Agpat2

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cytokine production
lipid metabolic process
phosphatidic acid biosynthetic process
metabolic process
phospholipid biosynthetic process
1-acylglycerol-3-phosphate O-acyltransferase activity
transferase activity
transferase activity, transferring acyl groups

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200026827.mm.1 NM_026212.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200026827.mm.1 TC0200003195.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200026823.mm.1 NM_026212.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200026823.mm.1 TC0200003195.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200026828.mm.1 NM_026212.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200026828.mm.1 TC0200003195.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200026819.mm.1 NM_026212.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200026819.mm.1 TC0200003195.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200026826.mm.1 NM_026212.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200026826.mm.1 TC0200003195.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200026824.mm.1 NM_026212.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200026824.mm.1 TC0200003195.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 349360 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0