This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01326904_m1
See other GPR107 GE Assays ›
Gene Symbol:
Gpr107
Gene Name:
G protein-coupled receptor 107
Gene Aliases:
9930033D15Rik, AI790205, C530034M11, mKIAA1624
Chromosome Location:
Chr.2: 31151997 - 31216567 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01326904_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr107
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178760.4 NP_848875.2 11 - 12 1054 71
GenBank mRNA AK030433.1 - 11 - 12 1050 71
AK034815.1 - 11 - 12 1037 71
AK082237.1 - 11 - 12 1052 71
AK083025.1 - 11 - 12 1048 71
AK088544.1 - 11 - 12 1038 71
AK129407.1 - 11 - 12 949 71
AK142454.1 - 11 - 12 1019 71
AK154530.1 - 11 - 12 1050 71
BC092231.1 - 11 - 12 1022 30607603 71
BC117900.1 - 11 - 12 1032 40097045 71
BC117901.1 - 11 - 12 1032 40097052 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000547.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr107

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200004547.mm.1 NM_178760.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200004547.mm.1 NM_178760.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200004547.mm.1 TC0200000547.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0