This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01326907_m1
See other GPR107 GE Assays ›
Gene Symbol:
Gpr107
Gene Name:
G protein-coupled receptor 107
Gene Aliases:
9930033D15Rik, AI790205, C530034M11, mKIAA1624
Chromosome Location:
Chr.2: 31151997 - 31216567 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01326907_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr107
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178760.4 NP_848875.2 15 - 16 1390 103
GenBank mRNA AK030433.1 - 15 - 16 1386 103
AK034815.1 - 15 - 16 1373 103
AK082237.1 - 15 - 16 1388 103
AK083025.1 - 15 - 16 1384 103
AK088544.1 - 15 - 16 1374 103
AK129407.1 - 15 - 16 1285 103
AK154530.1 - 15 - 16 1386 103
BC092231.1 - 15 - 16 1358 30607603 103
BC117900.1 - 15 - 16 1368 40097045 103
BC117901.1 - 15 - 16 1368 40097052 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000547.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr107

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200004560.mm.1 NM_178760.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200004560.mm.1 NM_178760.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200004560.mm.1 TC0200000547.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0