This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01327477_g1
See other PDIA4 GE Assays ›
Gene Symbol:
Pdia4
Gene Name:
protein disulfide isomerase associated 4
Gene Aliases:
AI987846, Cai, ERp-72, Erp72
Chromosome Location:
Chr.6: 47796141 - 47813512 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01327477_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pdia4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009787.2 NP_033917.2 5 - 6 1012 74
GenBank mRNA AK028292.1 - 5 - 6 862 74
AK146288.1 - 5 - 6 845 74
AK148801.1 - 5 - 6 934 74
AK150566.1 - 5 - 6 893 74
AK161534.1 - 5 - 6 850 74
AK168447.1 - 5 - 6 816 74
AK169387.1 - 5 - 6 857 74
BC066857.1 - 4 - 5 611 74
BC141078.1 - 5 - 6 908 9055690 74
J05186.1 - 5 - 6 895 74
Y00884.1 - 4 - 5 718 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002212.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pdia4

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
response to endoplasmic reticulum stress
cell redox homeostasis
chaperone-mediated protein folding
positive regulation of protein folding
protein disulfide isomerase activity
isomerase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600017383.mm.1 NM_009787.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600017383.mm.1 NM_009787.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600017383.mm.1 TC0600002212.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline