This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01327810_g1
See other CERCAM GE Assays ›
Gene Symbol:
Cercam
Gene Name:
cerebral endothelial cell adhesion molecule
Gene Aliases:
2600006K01Rik, AL024097, Ceecam1
Chromosome Location:
Chr.2: 29845105 - 29882845 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01327810_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cercam
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207298.2 NP_997181.1 7 - 8 995 57
GenBank mRNA AK155384.1 - 7 - 8 991 57
BC052176.1 - 7 - 8 956 57
BC138847.1 - 7 - 8 974 8861869 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000514.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cercam

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200003806.mm.1 NM_207298.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003806.mm.1 NM_207298.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200003806.mm.1 TC0200000514.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200003807.mm.1 NM_207298.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200003807.mm.1 NM_207298.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200003807.mm.1 TC0200000514.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline