This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01327860_m1
See other BFSP1 GE Assays ›
Gene Symbol:
Bfsp1
Gene Name:
beaded filament structural protein 1, in lens-CP94
Gene Aliases:
CP95
Chromosome Location:
Chr.2: 143826528 - 143915274 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01327860_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bfsp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291061.1 NP_001277990.1 3 - 4 751 68
NM_009751.2 NP_033881.2 3 - 4 751 68
GenBank mRNA AB003147.1 - 3 - 4 521 68
AK053766.1 - 3 - 4 522 68
AK143180.1 - 3 - 4 522 68
BC057580.1 - 3 - 4 751 6412634 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004749.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bfsp1

Gene Ontology Categories:

Function(s) Process(es)

cell maturation
lens fiber cell development
structural molecule activity
structural constituent of eye lens
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200040641.mm.1 NM_009751.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200040641.mm.1 NM_009751.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200040641.mm.1 TC0200004749.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0