This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01327928_g1
See other MEG3 GE Assays ›
Gene Symbol:
Meg3
Gene Name:
maternally expressed 3
Gene Aliases:
2900016C05Rik, 3110050O07Rik, 6330408G06Rik, AI425946, AW108224, D12Bwg1266e, Gtl2, R74756, R75394
Chromosome Location:
Chr.12: 109540996 - 109571729 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01327928_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Meg3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_003633.3 4 - 5 1467 55
NR_027651.2 8 - 9 1455 55
GenBank mRNA AK018105.1 - 4 - 5 1468 55
AK147321.1 - 6 - 7 2935 55
BC094421.1 - 8 - 9 1444 55
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CB202480.1 - 623 Chr. - - [-] on Build 55
GenBankmRNA BY610166.1 - 2 - 3 144 Chr. 12 - 109549757 - 109558923 [+] on Build GRCm38 55
GenBankmRNA CJ290411.1 - 2 - 3 210 Chr. 12 - 109549691 - 109558917 [+] on Build GRCm38 55
GenBankmRNA AV117970.1 - 2 - 3 213 Chr. 12 - 109549688 - 109558738 [+] on Build GRCm38 55
GenBankmRNA BY639887.1 - 2 - 3 156 Chr. 12 - 109549745 - 109558917 [+] on Build GRCm38 55
GenBankmRNA CF746141.1 - 5 - 6 484 Chr. 12 - 109544694 - 109558925 [+] on Build GRCm38 55
GenBankmRNA AV033589.2 - 4 - 5 465 Chr. 12 - 109546397 - 109559187 [+] on Build GRCm38 55
GenBankmRNA CX730605.1 - 2 - 3 142 Chr. 12 - 109549759 - 109559023 [+] on Build GRCm38 55
GenBankmRNA BQ954402.1 - 3 - 4 343 Chr. 12 - 109549476 - 109559105 [+] on Build GRCm38 55
GenBankmRNA BQ175478.1 - 1 - 2 Chr. 12 - 109549730 - 109559191 [-] on Build GRCm38 55
GenBankmRNA DN173243.1 - 1 - 2 103 Chr. 12 - 109558232 - 109559166 [+] on Build GRCm38 55
GenBankmRNA CF745386.1 - 3 - 4 287 Chr. 12 - 109549514 - 109559125 [+] on Build GRCm38 55
GenBankmRNA BY718623.1 - 4 - 5 471 Chr. 12 - 109546394 - 109558814 [+] on Build GRCm38 55
GenBankmRNA R75262.1 - 1 - 2 Chr. 12 - 109558281 - 109558882 [-] on Build GRCm38 55

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 1906-1 Mir1906-1 Mir1906,
Mirn1906,
mmu-mir-1906,
mmu-mir-1906-1
-13.6kb Sense Chr. 12 - 109544541 - 109544620 on NCBI Build GRCm38
Downstream microRNA 770 Mir770 Mirn770,
mir-770,
mmu-mir-770
4.6kb Sense Chr. 12 - 109563692 - 109563785 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s)

in utero embryonic development
liver development
DNA methylation
skeletal muscle tissue development
determination of adult lifespan
post-embryonic development
multicellular organism growth
lung alveolus development
SMAD protein signal transduction
genetic imprinting

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200008069.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008069.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008069.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008077.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008077.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008077.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008072.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008072.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008072.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008073.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008073.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008073.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008074.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008074.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008074.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008076.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008076.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008076.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008078.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008078.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008078.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008068.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008068.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008068.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008080.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008080.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008080.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008070.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008070.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008070.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008071.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008071.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008071.mm.1 TC1200001127.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200008079.mm.1 NR_003633.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200008079.mm.1 NR_027651.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200008079.mm.1 TC1200001127.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline