This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01327929_m1
See other NAPB GE Assays ›
Gene Symbol:
Napb
Gene Name:
N-ethylmaleimide sensitive fusion protein attachment protein beta
Gene Aliases:
Brp14, E161, I47, SNARE, b-SNAP
Chromosome Location:
Chr.2: 148693985 - 148732420 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01327929_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Napb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019632.3 NP_062606.1 1 - 2 153 86
GenBank mRNA AK078224.1 - 1 - 2 125 86
AK083184.1 - 1 - 2 154 86
BC038362.1 - 1 - 2 127 5368175 86
BC049874.1 - 1 - 2 200 5360432 86

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Napb

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
regulation of synaptic vesicle priming
protein transport
vesicle-mediated transport
synaptic transmission, glutamatergic
SNARE complex disassembly
membrane fusion
soluble NSF attachment protein activity
protein binding
syntaxin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z4mpm:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0