This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01328658_g1
See other CDAN1 GE Assays ›
Gene Symbol:
Cdan1
Gene Name:
congenital dyserythropoietic anemia, type I (human)
Gene Aliases:
1500015A01Rik, AI448026, AW492297, CDA-I, CDA1, CDAI
Chromosome Location:
Chr.2: 120716154 - 120731517 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01328658_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdan1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026891.2 NP_081167.2 20 - 21 2837 156
GenBank mRNA AK034144.1 - 5 - 6 530 156
AK157271.1 - 20 - 21 2838 156
BC117555.1 - 18 - 19 2122 40098039 156

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005501.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

chromatin organization
nuclear envelope organization
protein localization
negative regulation of DNA replication
chromatin assembly
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200037606.mm.1 NM_026891.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200037606.mm.1 NM_026891.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200037606.mm.1 TC0200005501.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline