This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01329405_m1
See other APBB1IP GE Assays ›
Gene Symbol:
Apbb1ip
Gene Name:
amyloid beta (A4) precursor protein-binding, family B, member 1 interacting protein
Gene Aliases:
9930118P07Rik, Prp48
Chromosome Location:
Chr.2: 22755989 - 22875653 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01329405_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apbb1ip
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019456.2 NP_062329.2 10 - 11 1361 81
GenBank mRNA AF020313.1 - 7 - 8 966 81
AK089987.1 - 2 - 3 235 81
BC003991.1 - 7 - 8 1012 81
BC023110.1 - 10 - 11 1250 3707478 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000352.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apbb1ip

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
signal transduction
positive regulation of cell adhesion
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200001721.mm.1 NM_019456.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200001721.mm.1 NM_019456.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200001721.mm.1 TC0200000352.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0