This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01329417_m1
See other YME1L1 GE Assays ›
Gene Symbol:
Yme1l1
Gene Name:
YME1-like 1 (S. cerevisiae)
Gene Aliases:
FtsH1, Ftsh
Chromosome Location:
Chr.2: 23156505 - 23199260 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01329417_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Yme1l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013771.5 NP_038799.1 15 - 16 1909 69
GenBank mRNA AF090430.1 - 15 - 16 1887 69
AK045141.1 - 15 - 16 1893 69
AK079175.1 - 15 - 16 1869 69
AY136286.1 - 15 - 16 1725 69
AY136287.1 - 15 - 16 1878 69
BC007128.1 - 15 - 16 1870 3591123 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000363.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Yme1l1

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
misfolded or incompletely synthesized protein catabolic process
mitochondrion organization
cell proliferation
nucleotide binding
metalloendopeptidase activity
ATP binding
peptidase activity
metallopeptidase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200001820.mm.1 NM_013771.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200001820.mm.1 NM_013771.5 Refseq
Affymetrix Mouse Clariom S Array PSR0200001820.mm.1 TC0200000363.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0