This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01329726_g1
See other ACP2 GE Assays ›
Gene Symbol:
Acp2
Gene Name:
acid phosphatase 2, lysosomal
Gene Aliases:
Acp-2, LAP
Chromosome Location:
Chr.2: 91202812 - 91214098 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01329726_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007387.2 NP_031413.1 2 - 3 231 54
GenBank mRNA AK089401.1 - 2 - 3 222 54
AK128963.1 - 2 - 3 224 54
AK133756.1 - 2 - 3 227 54
AK154269.1 - 2 - 3 227 54
AK170658.1 - 2 - 3 231 54
BC013495.1 - 2 - 3 221 54
BC023343.1 - 2 - 3 213 5136781 54
X57199.1 - 2 - 3 202 54

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001371.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acp2

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
lysosome organization
response to organic substance
dephosphorylation
autophagic cell death
phosphotyrosine binding
acid phosphatase activity
phosphoprotein phosphatase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200010578.mm.1 NM_007387.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200010578.mm.1 NM_007387.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200010578.mm.1 TC0200001371.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200010580.mm.1 NM_007387.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200010580.mm.1 NM_007387.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200010580.mm.1 TC0200001371.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0