This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01329728_g1
See other ACP2 GE Assays ›
Gene Symbol:
Acp2
Gene Name:
acid phosphatase 2, lysosomal
Gene Aliases:
Acp-2, LAP
Chromosome Location:
Chr.2: 91202812 - 91214098 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01329728_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007387.2 NP_031413.1 4 - 5 476 59
GenBank mRNA AK089401.1 - 4 - 5 467 59
AK128963.1 - 4 - 5 469 59
AK133756.1 - 4 - 5 472 59
AK154269.1 - 4 - 5 472 59
AK170658.1 - 4 - 5 476 59
BC013495.1 - 4 - 5 466 59
BC023343.1 - 4 - 5 458 5136781 59
X57199.1 - 4 - 5 447 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001371.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acp2

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
lysosome organization
response to organic substance
dephosphorylation
autophagic cell death
phosphotyrosine binding
acid phosphatase activity
phosphoprotein phosphatase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200010582.mm.1 NM_007387.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200010582.mm.1 NM_007387.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200010582.mm.1 TC0200001371.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200010583.mm.1 NM_007387.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200010583.mm.1 NM_007387.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200010583.mm.1 TC0200001371.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline