This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01329731_m1
See other ACP2 GE Assays ›
Gene Symbol:
Acp2
Gene Name:
acid phosphatase 2, lysosomal
Gene Aliases:
Acp-2, LAP
Chromosome Location:
Chr.2: 91202812 - 91214098 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01329731_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007387.2 NP_031413.1 7 - 8 799 82
GenBank mRNA AK089401.1 - 7 - 8 790 82
AK128963.1 - 7 - 8 792 82
AK154269.1 - 7 - 8 795 82
AK170658.1 - 7 - 8 799 82
BC013495.1 - 7 - 8 787 82
BC023343.1 - 7 - 8 781 5136781 82
X57199.1 - 7 - 8 770 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001371.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acp2

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
lysosome organization
response to organic substance
dephosphorylation
autophagic cell death
phosphotyrosine binding
acid phosphatase activity
phosphoprotein phosphatase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200010587.mm.1 NM_007387.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200010587.mm.1 NM_007387.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200010587.mm.1 TC0200001371.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline