This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01329732_g1
See other ACP2 GE Assays ›
Gene Symbol:
Acp2
Gene Name:
acid phosphatase 2, lysosomal
Gene Aliases:
Acp-2, LAP
Chromosome Location:
Chr.2: 91202812 - 91214098 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01329732_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007387.2 NP_031413.1 8 - 9 882 93
GenBank mRNA AK089401.1 - 8 - 9 873 93
AK128963.1 - 8 - 9 875 93
AK133756.1 - 8 - 9 878 93
AK154269.1 - 8 - 9 878 93
AK170658.1 - 8 - 9 882 93
BC013495.1 - 8 - 9 870 93
BC023343.1 - 8 - 9 864 5136781 93
X57199.1 - 8 - 9 853 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001371.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acp2

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
lysosome organization
response to organic substance
dephosphorylation
autophagic cell death
phosphotyrosine binding
acid phosphatase activity
phosphoprotein phosphatase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200010588.mm.1 NM_007387.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200010588.mm.1 NM_007387.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200010588.mm.1 TC0200001371.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline