This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01330637_m1
See other QSOX2 GE Assays ›
Gene Symbol:
Qsox2
Gene Name:
quiescin Q6 sulfhydryl oxidase 2
Gene Aliases:
BC030934, Qscn6l1, Soxn
Chromosome Location:
Chr.2: 26209124 - 26237399 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01330637_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Qsox2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK042036.1 - 6 - 7 848 80
AK140324.1 - 9 - 10 1226 80
AK157992.1 - 9 - 10 1219 80
AK165642.1 - 9 - 10 1228 80
BC030934.1 - 9 - 10 1260 4920915 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003182.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

oxidase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200026480.mm.1 NM_153559.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200026480.mm.1 TC0200003182.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline