This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01330696_g1
See other GPSM1 GE Assays ›
Gene Symbol:
Gpsm1
Gene Name:
G-protein signalling modulator 1 (AGS3-like, C. elegans)
Gene Aliases:
1810037C22Rik, AW107933, Ags3
Chromosome Location:
Chr.2: 26314606 - 26348237 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01330696_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpsm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199146.1 NP_001186075.1 11 - 12 1544 129
NM_001199147.1 NP_001186076.1 12 - 13 1793 129
NM_153410.5 NP_700459.2 12 - 13 1724 129
GenBank mRNA AK048691.1 - 12 - 13 1718 129
AK075789.1 - 2 - 3 297 129
AK082985.1 - 12 - 13 1668 129
AK084631.1 - 11 - 12 1486 129
AK177427.1 - 2 - 3 173 129
BC027004.1 - 9 - 10 1075 129
BC071197.1 - 12 - 13 1790 6465025 129
L23316.1 - 3 - 4 351 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000445.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpsm1

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
nervous system development
regulation of G-protein coupled receptor protein signaling pathway
cell differentiation
G-protein alpha-subunit binding
GDP-dissociation inhibitor activity
protein binding
GTPase regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200002814.mm.1 NM_001199146.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002814.mm.1 NM_001199146.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002814.mm.1 NM_001199147.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002814.mm.1 NM_001199147.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002814.mm.1 NM_153410.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002814.mm.1 NM_153410.5 Refseq
Affymetrix Mouse Clariom S Array PSR0200002814.mm.1 TC0200000445.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200002815.mm.1 NM_001199146.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002815.mm.1 NM_001199146.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002815.mm.1 NM_001199147.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002815.mm.1 NM_001199147.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002815.mm.1 NM_153410.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002815.mm.1 NM_153410.5 Refseq
Affymetrix Mouse Clariom S Array PSR0200002815.mm.1 TC0200000445.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0