This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01330701_m1
See other GPSM1 GE Assays ›
Gene Symbol:
Gpsm1
Gene Name:
G-protein signalling modulator 1 (AGS3-like, C. elegans)
Gene Aliases:
1810037C22Rik, AW107933, Ags3
Chromosome Location:
Chr.2: 26314606 - 26348237 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01330701_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpsm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199146.1 NP_001186075.1 8 - 9 1195 65
NM_001199147.1 NP_001186076.1 8 - 9 1264 65
NM_153410.5 NP_700459.2 8 - 9 1195 65
GenBank mRNA AK048691.1 - 8 - 9 1189 65
AK082985.1 - 8 - 9 1139 65
AK084631.1 - 8 - 9 1137 65
BC027004.1 - 5 - 6 546 65
BC071197.1 - 8 - 9 1261 6465025 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000445.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpsm1

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
nervous system development
regulation of G-protein coupled receptor protein signaling pathway
cell differentiation
G-protein alpha-subunit binding
GDP-dissociation inhibitor activity
protein binding
GTPase regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200002800.mm.1 NM_001199146.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002800.mm.1 NM_001199146.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002800.mm.1 NM_001199147.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002800.mm.1 NM_001199147.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002800.mm.1 NM_153410.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200002800.mm.1 NM_153410.5 Refseq
Affymetrix Mouse Clariom S Array PSR0200002800.mm.1 TC0200000445.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0