This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01331615_m1
See other NAPB GE Assays ›
Gene Symbol:
Napb
Gene Name:
N-ethylmaleimide sensitive fusion protein attachment protein beta
Gene Aliases:
Brp14, E161, I47, SNARE, b-SNAP
Chromosome Location:
Chr.2: 148693985 - 148732420 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01331615_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Napb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019632.3 NP_062606.1 3 - 4 349 69
GenBank mRNA AK083184.1 - 3 - 4 350 69
BC038362.1 - 3 - 4 323 5368175 69
BC049874.1 - 3 - 4 396 5360432 69
X61455.1 - 2 - 3 140 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Napb

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
regulation of synaptic vesicle priming
protein transport
vesicle-mediated transport
synaptic transmission, glutamatergic
SNARE complex disassembly
membrane fusion
soluble NSF attachment protein activity
protein binding
syntaxin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline