This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01331645_g1
See other PLD4 GE Assays ›
Gene Symbol:
Pld4
Gene Name:
phospholipase D family, member 4
Gene Aliases:
AI132321, thss
Chromosome Location:
Chr.12: 112760655 - 112768988 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01331645_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pld4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178911.4 NP_849242.1 10 - 11 1422 73
GenBank mRNA AK079549.1 - 10 - 11 1381 73
AK079878.1 - 10 - 11 1424 73
AK151551.1 - 10 - 11 1656 73
AK152969.1 - 9 - 10 1271 73
BC058565.1 - 10 - 11 1372 6442038 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001272.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pld4

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
lipid metabolic process
phagocytosis
lipid catabolic process
catalytic activity
phospholipase D activity
protein binding
hydrolase activity
N-acylphosphatidylethanolamine-specific phospholipase D activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK170362.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 BC058565.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK152969.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK079878.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK151551.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK170362.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 BC058565.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK152969.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK079878.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591153 AK151551.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261375 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132916 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244269 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544148 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 95131 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158352 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK170362.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 BC058565.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK152969.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK079878.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK151551.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK170362.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 BC058565.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK152969.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK079878.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591153 AK151551.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752292 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752290 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752294 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752313 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752310 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752333 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752323 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752319 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752300 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752314 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752303 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752316 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752320 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752331 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752298 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752334 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752329 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752328 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752326 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752296 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752311 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752321 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752325 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28752302 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158352 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07755088 - -
Whole Mouse Genome G4122A A_52_P591153 AK170362.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P591153 BC058565.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P591153 AK152969.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P591153 AK079878.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P591153 AK151551.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P591153 AK170362.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P591153 BC058565.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P591153 AK152969.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P591153 AK079878.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P591153 AK151551.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115714 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158352 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09963115 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09877961 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10304230 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10304250 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10304240 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752425 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752356 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752411 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752389 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752386 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752397 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752300 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752292 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752359 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752413 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752320 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752383 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752329 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752323 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752280 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752414 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752352 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752331 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752408 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752384 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752371 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752427 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752422 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752416 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752349 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752311 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752400 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752350 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752418 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752296 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752346 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752302 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752325 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752367 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752283 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752387 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752420 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752407 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752373 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752395 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752340 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752326 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752394 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752405 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752355 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752290 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752319 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752385 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752343 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752365 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752278 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752402 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752375 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752392 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752321 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752348 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752282 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752281 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752424 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752316 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752363 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28752328 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752350 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752302 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752316 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752356 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752354 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10399537 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752349 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752346 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752383 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752303 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752319 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752345 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752379 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752365 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752339 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752320 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752375 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752359 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752323 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752290 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752285 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10374915 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752382 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752296 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752371 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752310 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752348 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752309 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752361 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P09979472 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752355 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10304243 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P09979466 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752369 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10399555 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752343 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752331 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752328 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752321 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752311 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752286 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752368 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P09899983 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752313 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752340 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10402905 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752292 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752341 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752300 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752381 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752294 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P09979460 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752314 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10402904 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752334 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752329 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752377 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P09979475 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10304236 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10399541 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10304246 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P28752357 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10398910 - -
Affymetrix Mouse Gene 1.0 ST Array 10398909 - -
Affymetrix Mouse Gene 1.0 ST Array 10398918 - -
Affymetrix Mouse Gene 1.0 ST Array 10398912 - -
Affymetrix Mouse Gene 1.0 ST Array 10398911 - -
Affymetrix Mouse Gene 1.0 ST Array 10398919 - -
Affymetrix Mouse Gene 1.0 ST Array 10398915 - -
Affymetrix Mouse Gene 1.0 ST Array 10398916 - -
Affymetrix Mouse Gene 1.0 ST Array 10398914 - -
Affymetrix Mouse Gene 1.0 ST Array 10398908 - -
Affymetrix Mouse Gene 1.0 ST Array 10398917 - -
Affymetrix Mouse Gene 1.0 ST Array 10398913 - -
Affymetrix HT MG-430 PM Array Plate 1433678_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10398907 - -
Mouse Genome 430 2.0 Array 1433678_PM_at - -
Mouse Genome 430 2.0 Array 1433678_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 104759_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 104759_at - -
Affymetrix Mouse Clariom S Array PSR1200009163.mm.1 NM_178911.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200009163.mm.1 NM_178911.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200009163.mm.1 TC1200001272.mm.1 MTA TC
Mouse Expression Set 430 1433678_PM_at - -
Mouse Expression Set 430 1433678_at - -
Murine Genome U74v2 Set 103299_at - -
Murine Genome U74v2 Set 162043_PM_i_at - -
Murine Genome U74v2 Set 103299_PM_at - -
Murine Genome U74v2 Set 162043_i_at - -
Murine Genome U74v2 Set 168401_PM_f_at - -
Murine Genome U74v2 Set 168401_f_at - -
Mouse Genome 430A 2.0 Array 1433678_at - -
Mouse Genome 430A 2.0 Array 1433678_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1433678_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1433678_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 6590066 - -
Illumina mouse-6 v1.0 expression beadchip 6590066 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006590066 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234223 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1234223 - -
mouse-6 v1.1 (Illumina) scl42725.12.1_79-S - -
Illumina Sentrix mouse-6.v1 bead array 6590066 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl42725.12.1_79-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6590066 - -
MouseRef-8 v2 Expression BeadChip ILMN_1234223 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234223 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-t8jrk:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0