This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01331665_g1
See other BRF1 GE Assays ›
Gene Symbol:
Brf1
Gene Name:
BRF1 homolog, subunit of RNA polymerase III transcription initiation factor IIIB (S. cerevisiae)
Gene Aliases:
2510002F24Rik, GTF3B, TAF3C, TAFIII90, TFIIIB90, mTFIIIB90
Chromosome Location:
Chr.12: 112955631 - 113000707 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01331665_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Brf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028193.3 NP_082469.2 9 - 10 1221 99
GenBank mRNA CF737042.1 - 6 - 7 542 99

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Brf1

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase III promoter
regulation of mRNA stability
positive regulation of transcription from RNA polymerase III promoter
molecular_function
zinc ion binding
TBP-class protein binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0