This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01332249_m1
See other GTF2H5 GE Assays ›
Gene Symbol:
Gtf2h5
Gene Name:
general transcription factor IIH, polypeptide 5
Gene Aliases:
2700017P07Rik, 2810432H05Rik, D17Wsu155e
Chromosome Location:
Chr.17: 6079828 - 6085488 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01332249_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2h5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181392.3 NP_852057.2 2 - 3 163 77
GenBank mRNA AK003547.1 - 2 - 3 152 77
AK003579.1 - 2 - 3 147 77
AK011281.1 - 2 - 3 148 77
AK012257.1 - 2 - 3 225 77
AK019011.1 - 2 - 3 118 77
BC029238.1 - 2 - 3 139 4489531 77
W50582.1 - 2 - 3 162 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK003547.1, AK003579.1, AK011281.1, AK012257.1, BC029238.1, NM_181392.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf2h5

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
nucleotide-excision repair
nucleotide-excision repair, preincision complex assembly
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription elongation from RNA polymerase I promoter
rRNA processing
cellular response to DNA damage stimulus
cellular response to gamma radiation
rDNA binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31020950 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31020949 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31020951 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022404 NM_181392.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022404 AK011281.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022404 AK012257.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022404 AK013225.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566923 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145471 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768635 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198427 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42601 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128431 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022404 NM_181392.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022404 AK011281.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022404 AK012257.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022404 AK013225.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020974 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020976 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020951 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020983 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020950 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020957 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020960 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020961 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020958 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020972 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020964 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020962 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020979 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020970 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020955 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020952 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020954 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31020981 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128431 - -
Whole Mouse Genome G4122A A_51_P198271 AK084443.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P230778 NM_181392.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P230778 AK003547.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P230778 AK003579.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P230778 AK011281.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P230778 AK012257.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118324 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128431 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13441059 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13216241 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31020951 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31020950 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31020949 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020957 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020977 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021006 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020958 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021032 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020986 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020993 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021024 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021018 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021014 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021004 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020948 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020970 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021013 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021002 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020955 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020972 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020995 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021027 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020974 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020968 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021040 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020976 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021008 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021000 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020982 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021017 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020984 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020961 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021022 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021037 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021010 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020952 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020987 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020979 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020981 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021038 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020992 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020954 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021035 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021033 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020951 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021021 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021025 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021031 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021029 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31020988 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31021039 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31021013 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020995 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13157743 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31021010 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020974 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020986 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020969 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020962 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020958 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020981 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31021008 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13333840 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020988 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13255488 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020992 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020952 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020957 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31021017 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020987 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31021014 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020951 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31021006 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020968 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020961 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020970 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020977 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020993 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020955 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020948 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020982 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31021004 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020972 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020964 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020994 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020954 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020976 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31020984 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10441492 - -
Affymetrix Mouse Gene 1.0 ST Array 10441491 - -
Affymetrix Mouse Gene 1.0 ST Array 10441493 - -
Affymetrix Mouse Gene 1.0 ST Array 10441490 - -
Affymetrix HT MG-430 PM Array Plate 1452955_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1428906_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10441489 - -
Mouse Genome 430 2.0 Array 1452955_at - -
Mouse Genome 430 2.0 Array 1428906_at - -
Mouse Genome 430 2.0 Array 1452955_PM_at - -
Mouse Genome 430 2.0 Array 1428906_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66467_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66467_at - -
Affymetrix Mouse Clariom S Array PSR1700000314.mm.1 NM_181392.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700000314.mm.1 NM_181392.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700000314.mm.1 TC1700000048.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700000313.mm.1 NM_181392.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700000313.mm.1 NM_181392.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700000313.mm.1 TC1700000048.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700000315.mm.1 NM_181392.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700000315.mm.1 NM_181392.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700000315.mm.1 TC1700000048.mm.1 MTA TC
Mouse Expression Set 430 1452955_PM_at - -
Mouse Expression Set 430 1452955_at - -
Mouse Expression Set 430 1428906_at - -
Mouse Expression Set 430 1428906_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1452955_at - -
Affymetrix HT Mouse Genome 430B Array 1428906_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1428906_at - -
Affymetrix HT Mouse Genome 430B Array 1452955_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 452215 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 2510563 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002510563 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223234 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1223234 - -
mouse-6 v1.1 (Illumina) scl50382.4_247-S - -
Illumina Sentrix mouse-6.v1 bead array 2510563 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl50382.4_247-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2510563 - -
MouseRef-8 v2 Expression BeadChip ILMN_1223234 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223234 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50382.4_247-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline