This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01332259_m1
See other PAK3 GE Assays ›
Gene Symbol:
Pak3
Gene Name:
p21 protein (Cdc42/Rac)-activated kinase 3
Gene Aliases:
PAK-3, Pak65alpha, Pak65beta, Stk4
Chromosome Location:
Chr.X: 143518368 - 143797796 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01332259_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pak3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195049.1 NP_001181978.1 4 - 5 478 135
GenBank mRNA U39738.1 - 4 - 5 137

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001195049.1, U39738.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Protein Kinase Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
activation of MAPK activity
protein phosphorylation
Rho protein signal transduction
multicellular organism development
regulation of mitotic cell cycle
axonogenesis
metabolic process
regulation of neuron projection development
phosphorylation
dendrite development
cell migration
signal transduction by protein phosphorylation
actin cytoskeleton organization
regulation of actin filament polymerization
stress-activated protein kinase signaling cascade
activation of protein kinase activity
regulation of apoptotic process
regulation of MAPK cascade
positive regulation of neuron apoptotic process
dendritic spine development
positive regulation of dendritic spine morphogenesis
cellular response to organic cyclic compound
positive regulation of DNA biosynthetic process
nucleotide binding
catalytic activity
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
Rho GTPase binding
SH3 domain binding
metal ion binding
Rac GTPase binding
GTPase binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00008959.mm.1 NM_001195046.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008959.mm.1 NM_001195046.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008959.mm.1 NM_001195047.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008959.mm.1 NM_001195047.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008959.mm.1 NM_001195048.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008959.mm.1 NM_001195048.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008959.mm.1 NM_001195049.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008959.mm.1 NM_001195049.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008959.mm.1 NM_008778.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008959.mm.1 NM_008778.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008959.mm.1 TC0X00001413.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00008960.mm.1 NM_001195046.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008960.mm.1 NM_001195046.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008960.mm.1 NM_001195047.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008960.mm.1 NM_001195047.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008960.mm.1 NM_001195048.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008960.mm.1 NM_001195048.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008960.mm.1 NM_001195049.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008960.mm.1 NM_001195049.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008960.mm.1 NM_008778.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008960.mm.1 NM_008778.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00008960.mm.1 TC0X00001413.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 676546 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0